missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N), Honeywell Fluka™
12.71 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 L
5 L
10 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Linear Formula | NaOH |
Concentration | 0.1M |
UN Number | NONH for all modes of transport |
Quantity | 1 L |
Formula Weight | 40g/mol |
Packaging | Plastic Bottle |
Vapor Pressure | 3 mmHg (37°C) |
Chemical Name or Material | Sodium hydroxide solution |
Safety and Handling
Product Identifier
- Sodium hydroxide solution
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
- MIXTURE LIST-Contains : sodium hydroxide; caustic soda
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'