missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, 0.1M, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
6 x 500 mL
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Linear Formula | NaOH |
Concentration | 0.1M |
UN Number | NONH for all modes of transport |
Density | 1g/mL at 25°C |
Quantity | 2 L |
Formula Weight | 40g/mol |
Vapor Density | >1 (vs air) |
Vapor Pressure | 3mmHg (37°C) |
Chemical Name or Material | Sodium Hydroxide Solution |
Safety and Handling
Product Identifier
- Sodium hydroxide solution
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
- MIXTURE LIST-Contains : sodium hydroxide; caustic soda
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'