missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. 10131860
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy
This item is not returnable. View return policy

HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide solution, sodium hydroxide na oh, sodium hydrate, soda, caustic, white caustic, soda lye, aetznatron, caustic soda, ascarite, 1L, CHEBI:32145, Colorless, 39.99g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 13
Color Colorless
Quantity 10 L
Formula Weight 39.99g/mol
Packaging Bag in box
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide, 0.2 N standard solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'