missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
£58.80 - £435.00
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | 3548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10488790
|
Fisher Chemical
J/7800/15 |
1 L | HDPE plastic bottle |
£58.80
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10040220
|
Fisher Chemical
J/7800/17 |
2.5 L | HDPE plastic bottle |
£111.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306240
|
Fisher Chemical
J/7800/21 |
5 L | HDPE plastic bottle |
£138.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10715721
|
Fisher Chemical
J/7800/24 |
10 L | Bag in box |
£128.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10498980
|
Fisher Chemical
J/7800/25 |
25 L | Plastic container |
£435.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
3548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
12 | |
1310-73-2 | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 |
120°C | |
8°C | |
3548 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
Liquid |