Learn More
Sodium Hydroxide Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
HNaO, CAS Number-1310-73-2, white caustic, sodium hydroxide solution, sodium hydrate, sodium hydroxide, soda, caustic, soda lye, ascarite, caustic soda, aetznatron, sodium hydroxide na oh, 6 ampules, >100 deg.C, CHEBI:32145, Undesignated, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, -10 deg.C
Brand: Fisher Chemical J/7620C/90
108.40 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 23
Additional Details : CAS Number : 1310-73-2 Weight : 0.94000kg Transport : UN number : 1824 Chem class : 8 Pack group : II
Description
HNaO, CAS Number-1310-73-2, white caustic, sodium hydroxide solution, sodium hydrate, sodium hydroxide, soda, caustic, soda lye, ascarite, caustic soda, aetznatron, sodium hydroxide na oh, 6 ampules, >100 deg.C, CHEBI:32145, Undesignated, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, -10 deg.C
Specifications
12 | |
1310-73-2 | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
Ampoule | |
14mmHg |
>100°C | |
-10°C | |
3548 | |
6 Ampules | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
> 1.0 | |
Liquid |
Safety and Handling
- Sodium hydroxide 1M solution (10M concentrate ampoules)
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide