missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N), Honeywell Fluka™
Volumetric, 0.5 M NaOH (0.5N)
£20.02 - £210.05
Chemical Identifiers
CAS | 1310-73-2, 7732-18-5 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15614900
|
Honeywell-Fluka
35257-1L |
1 L | Poly Bottle |
£20.02
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15644900
|
Honeywell-Fluka
35257-5L-VP |
5 L | Cubitainer™ | N/A | |||||
15634900
|
Honeywell-Fluka
35257-5L |
5 L | Poly Bottle |
£69.69
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15654900
|
Honeywell-Fluka
35257-6X1L |
6 x 1 L | Poly Bottle |
£127.72
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15604900
|
Honeywell Fluka
35257-10L-VP |
10 L | Cubitainer™ |
£210.05
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15694890
|
Honeywell-Fluka
35257-10L |
10 L | Poly Bottle | N/A | |||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
1310-73-2, 7732-18-5 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
>100°C | |
≥90.0000%, ≥1.0000% | |
HNaO | |
0.5 M NaOH (0.5N) | |
UN1824 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Volumetric |
1310-73-2, 7732-18-5 | |
≤100.0000%, <2.0000% | |
NaOH | |
MFCD00003548 | |
1.020 g/cm3 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40.00 | |
Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N) |
Safety and Handling
missing translation for 'shelfLife' : 450 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Sodium hydroxide solution
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide; caustic soda
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title