missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Volumetric, 2 M NaOH (2N), Honeywell Fluka™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Poly Bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 105°C to 140°C |
CAS Min % | ≥90.0000%, ≥5.0000% |
CAS Max % | ≤100.0000%, <10.0000% |
Linear Formula | NaOH |
Concentration | 2 M NaOH (2N) |
UN Number | UN1824 |
Density | 1.08 g/cm3 |
Quantity | 1 L |
Formula Weight | 40.0 |
Grade | Volumetric |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- sodium hydroxide solution
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium hydroxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'