Learn More
Sodium hypochlorite, 10-15% active chlorine, Thermo Scientific Chemicals
£22.81 - £175.11
Chemical Identifiers
CAS | 7681-52-9, 7732-18-5 |
---|---|
Molecular Formula | ClNaO |
Molecular Weight (g/mol) | 74.44 |
MDL Number | MFCD00011120 |
InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
Synonym | sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution |
PubChem CID | 23665760 |
ChEBI | CHEBI:32146 |
IUPAC Name | sodium;hypochlorite |
SMILES | [O-]Cl.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10401841
|
Thermo Scientific Acros
219255000 |
500 mL | Plastic bottle |
£22.81
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10296650
|
Thermo Scientific Acros
219250025 |
2.5 L | Plastic bottle |
£60.78
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10553864
|
Thermo Scientific Acros
219250100 |
10 L | Plastic drum |
£175.11
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7681-52-9, 7732-18-5 | |
74.44 | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
23665760 | |
sodium;hypochlorite |
ClNaO | |
MFCD00011120 | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
CHEBI:32146 | |
[O-]Cl.[Na+] |
Specifications
-16.0°C | |
Yellow | |
ClNaO | |
MFCD00011120 | |
15,8762 | |
Solubility in water: soluble | |
[O-]Cl.[Na+] | |
74.44 | |
CHEBI:32146 | |
≥12% | |
1.2000g/mL |
7681-52-9, 7732-18-5 | |
Solution | |
NaOCl | |
01,1084; 04,456; 05,617; 06,543; 07,337; 08,461; 09,430; 10,365; 11,487; 15,293; 16,308 | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
sodium;hypochlorite | |
23665760 | |
74.44 | |
1.2 | |
Sodium hypochlorite, 13% Active chlorine |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting effects.
Contact with acids liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-668-3
missing translation for 'rtecsNumber' : NH3486300
missing translation for 'tsca' : TSCA
- Sodium hypochlorite
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium hypochlorite
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.