missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite solution Strong EU Pharmacopoeia, Fisher Chemical™

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Brand:  Fisher Chemical RP 203 1000

Code : 04

Additional Details : Weight : 1.00000kg Transport : UN number : 1791 Chem class : 8 Pack group : III


Product Code. 12910544

  • £111.00 / 1000mL
Estimated Shipment: 02-08-2024
to see stock.

Description

Description

Specifications

Specifications

Bottle
EU Ph Ref: 1081600
Sodium Hypochlorite
1000 mL
Eu. Ph.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium hypochlorite solution,strong. European Pharmacopoeia Ref: 1081600 (RP 203)

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash hands, forarms and face thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.

Supplemental information
  • MIXTURE LIST-Contains : sodium hypochlorite

SDS
Documents

Documents

Certificates
Special Offers

Special Offers