missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hypophosphite monohydrate, Puriss. p.a., Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
1 kg
50 kg
Specifications
Spezifikation
Quantity | 1 kg |
Linear Formula | NaH2PO2 · H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 105.99g/mol |
Percent Purity | ≥99% |
Grade | puriss. p.a. |
Chemical Name or Material | Sodium Hypophosphite Monohydrate, Reag. Ph. Eur. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'