missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hypophosphite monohydrate, Puriss. p.a., Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
1 kg
50 kg
Specifications
CAS Max % | 100.0000% |
Quantity | 50 kg |
Linear Formula | NH4F |
UN Number | NONH for all modes of transport |
Formula Weight | 105.99 |
Percent Purity | >99% |
Grade | puriss. p.a. |
Packaging | Fiberboard Box |
Chemical Name or Material | Sodium hypophosphite monohydrate, Puriss. p.a., Reag. Ph. Eur., ≥99% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'