missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hypophosphite monohydrate, Thermo Scientific Chemicals
£27.90 - £65.20
Specifications
CAS | 10039-56-2 |
---|---|
Physical Form | Solid |
Molecular Formula | NaH{2}PO{2}^.H{2}O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307707
|
Thermo Scientific Alfa Aesar
014104.30 |
250 g |
£27.90
250g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11317707
|
Thermo Scientific Alfa Aesar
014104.A1 |
1 kg |
£65.20
1kg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sodium hypophosphite monohydrate is used for electroless nickel plating agent and water treatment agent. It is also used in petroleum fields. Further, it is used as a food additive. In addition, it reduces nickel ions in solution to metallic nickel on metal substrates as well as on plastic substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
10039-56-2 | |
Solid | |
NaH{2}PO{2}^.H{2}O |