Learn More
Sodium ingot, 99.8% (metals basis), Thermo Scientific Chemicals
£104.00 - £387.00
Chemical Identifiers
CAS | 7440-23-5 |
---|---|
Molecular Formula | Na |
Molecular Weight (g/mol) | 22.99 |
MDL Number | MFCD00085307 |
InChI Key | KEAYESYHFKHZAL-UHFFFAOYSA-N |
Synonym | natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot |
PubChem CID | 5360545 |
ChEBI | CHEBI:26708 |
IUPAC Name | sodium |
SMILES | [Na] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341428
|
Thermo Scientific Alfa Aesar
010342.35 |
450 g |
£104.00
450g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355778
|
Thermo Scientific Alfa Aesar
010342.60 |
5 x 450 g |
£387.00
2250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-23-5 | |
22.99 | |
KEAYESYHFKHZAL-UHFFFAOYSA-N | |
5360545 | |
sodium |
Na | |
MFCD00085307 | |
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot | |
CHEBI:26708 | |
[Na] |
Specifications
Silver to Gray | |
Silver Gray ingot | |
99.8% (Metals basis) | |
MFCD00085307 | |
KEAYESYHFKHZAL-UHFFFAOYSA-N | |
sodium | |
5360545 | |
ACS | |
1.59 hPa (1mm Hg) | |
Sodium, Product meets ACS specifications |
7440-23-5 | |
0.968g/cm3 | |
Na | |
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot | |
[Na] | |
22.99 | |
CHEBI:26708 | |
Odorless | |
Ingot |
Safety and Handling
P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H260-H314-H335
missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: I
missing translation for 'einecsNumber' : 231-132-9
missing translation for 'rtecsNumber' : VY0686000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only