missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide, 99+%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10689253
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
10 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1300.0°C
Melting Point 661.0°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99% min. (on dried substance) (Argentometry)
Linear Formula NaI
Fieser 01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 10,365; 11,488; 12,29; 13,43; 14,89; 16,86
Merck Index 15,8765
Solubility Information Solubility in water: 184g/100mL (25°C). Other solubilities: soluble in alcohol,glycerol and acetone
Formula Weight 149.89
Percent Purity ≥99%
Grade Pure
Loss on Drying 3% max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
Specific Gravity 3.66
Density 3.6600g/mL
Chemical Name or Material Sodium iodide, Anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium iodide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'