missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Boiling Point | 1300°C |
Melting Point | 661°C |
Color | White |
pH | 6 to 9 |
Physical Form | Powder/Solid |
Quantity | 100 g |
Formula Weight | 149.89g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1.3mbar 767 |
Safety and Handling
Product Identifier
- Sodium iodide
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'