missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium iodide, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 g |
Linear Formula | NaI |
UN Number | UN3077 |
Formula Weight | 149.89g/mol |
Percent Purity | ≥99.5% |
Density | Vapor Density: >1 (vs air) |
Chemical Name or Material | Sodium iodide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'