missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide (White Crystals/Molecular Biology), Fisher BioReagents

Product Code. 10439503
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
This sodium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1300°C
Melting Point 661°C
CAS 7681-82-5
Color White
pH 6 to 9
Physical Form Powder/Solid
Quantity 100 g
Assay Percent Range ≥99 %
Molecular Formula INa
Merck Index 15, 8765
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES [Na+].[I-]
IUPAC Name sodium;iodide
Molecular Weight (g/mol) 149.894
PubChem CID 5238
ChEBI CHEBI:33167
Formula Weight 149.89
Percent Purity ≥99%
Grade Molecular Biology
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 1.3mbar at 767°C
Density 3.660g/cm³
Chemical Name or Material Sodium Iodide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium iodide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'