missing translation for 'onlineSavingsMsg'
Learn More

Sodium isopropoxide, Thermo Scientific Chemicals

Catalog Number 11483770
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
683-60-3
C3H7NaO
82.078
MFCD00061566
WBQTXTBONIWRGK-UHFFFAOYSA-N
sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate
10920443
sodium;propan-2-olate
CC(C)[O-].[Na+]
This item is not returnable. View return policy
683-60-3
C3H7NaO
82.078
MFCD00061566
WBQTXTBONIWRGK-UHFFFAOYSA-N
sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate
10920443
sodium;propan-2-olate
CC(C)[O-].[Na+]

Sodium isopropoxide is used in the preparation of ibuprofen intermediate. It is involved in reactions such as polymerization of butadiene. Further, it acts as a catalyst in the manufacturing of acetone and its derivatives. In addition, it is used as a solvent and serves as an antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70°C to 75°C
Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble in tetrahydrofuran.
Formula Weight 82.08
Physical Form Crystalline
Chemical Name or Material Sodium isopropoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'