missing translation for 'onlineSavingsMsg'
Learn More
Sodium L-lactate, 98+%, Thermo Scientific Chemicals Product Code.: 11363556

Sodium L-lactate, 98+%, Thermo Scientific Chemicals

Product Code. 11363556
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11363556

Brand: Thermo Scientific Alfa Aesar L14500.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium L-lactate is used as a food additive, preservative, acidity regulator and bulking agent. It finds application in shampoo and liquid soaps and other related products. It acts as an effective humectant and moisturizer. It is used in the treatment of arrhythmias.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium L-lactate is used as a food additive, preservative, acidity regulator and bulking agent. It finds application in shampoo and liquid soaps and other related products. It acts as an effective humectant and moisturizer. It is used in the treatment of arrhythmias.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-56-1
Molecular Formula C3H5NaO3
MDL Number MFCD00066576
Synonym sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium saltShow MoreShow Less

Specifications

Melting Point ∼160°C
Quantity 25 g
Beilstein 4567087
Solubility Information Soluble in water.
Formula Weight 112.06
Percent Purity ≥98%
Sensitivity Hygroscopic
Chemical Name or Material Sodium L-lactate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10234790 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberL14500.14,L14500.06,L14500.PG
Lot Number10201409 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14500.14,L14500.06
Lot Number10178165 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14500.14,L14500.06
Lot Number10193762 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14500.14,L14500.06
Lot Number10211596 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14500.14,L14500.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium L-lactate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.