missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium meta-Periodate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
£676.00 - £1905.00
Chemical Identifiers
CAS | 7790-28-5 |
---|---|
Molecular Formula | INaO4 |
Molecular Weight (g/mol) | 213.89 |
MDL Number | MFCD00003534 |
InChI Key | JQWHASGSAFIOCM-UHFFFAOYSA-M |
Synonym | sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate |
PubChem CID | 23667635 |
ChEBI | CHEBI:75226 |
SMILES | [Na+].[O-][I](=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15172017
|
Avantor J.T.Baker
3756-04 |
125 g | Poly Bottle |
£676.00
125g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15568164
|
Avantor J.T.Baker
3756-01 |
500 g | Poly Bottle |
£1905.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7790-28-5 | |
213.89 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
23667635 | |
[Na+].[O-][I](=O)(=O)=O |
INaO4 | |
MFCD00003534 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
CHEBI:75226 |
Specifications
7790-28-5 | |
MFCD00003534 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
sodium periodate | |
23667635 | |
213.89 | |
1L = 3.87kg |
INaO4 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
[Na+].[O-][I](=O)(=O)=O | |
213.89 | |
CHEBI:75226 | |
Meets ACS reagent chemical requirements | |
Sodium meta-Periodate |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title