missing translation for 'onlineSavingsMsg'
Learn More

Sodium metaperiodate, 98%, Thermo Scientific Chemicals

Product Code. 11341339
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Source of Periodic Acid, As An Analytical Reagent, and As An Oxidizing Agent

Source of periodic acid, as an analytical reagent, and as an oxidizing agent Sodium metaperiodate is used as an oxidizing reagent involved in the oxidation of cellulose. It is involved in the cleavage of vicinal diols to prepare two aldehydes. This oxidation is commonly used in selectively label RNA, saccharides and other compounds with vicinal diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
IUPAC Name sodium;periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 300°C (decomposition)
Color Colorless
Physical Form Powder
Quantity 2 kg
Assay Percent Range 98%
Solubility Information Soluble in water,sulfuric acid,nitric acid and acetic acid.
Formula Weight 213.89
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Appearance Colorless powder
Density 3.86
Chemical Name or Material Sodium metaperiodate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'