Learn More
Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Maximum level of impurities: Other halogens (as Cl) 0.02%, Mn 3ppm
£80.60 - £885.00
Chemical Identifiers
CAS | 7790-28-5 |
---|---|
Molecular Formula | INaO4 |
Molecular Weight (g/mol) | 213.89 |
MDL Number | MFCD00003534 |
InChI Key | JQWHASGSAFIOCM-UHFFFAOYSA-M |
Synonym | sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate |
PubChem CID | 23667635 |
ChEBI | CHEBI:75226 |
IUPAC Name | sodium;periodate |
SMILES | [Na+].[O-][I](=O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458400
|
Thermo Scientific Alfa Aesar
044309.22 |
100 g |
£80.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448400
|
Thermo Scientific Alfa Aesar
044309.36 |
500 g |
£284.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438400
|
Thermo Scientific Alfa Aesar
044309.A3 |
2 kg |
£885.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium metaperiodate is used as an oxidizing reagent involved in the oxidation of cellulose. It is involved in the cleavage of vicinal diols to prepare two aldehydes. This oxidation is commonly used in selectively label RNA, saccharides and other compounds with vicinal diols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-28-5 | |
213.89 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
23667635 | |
sodium;periodate |
INaO4 | |
MFCD00003534 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
CHEBI:75226 | |
[Na+].[O-][I](=O)(=O)=O |
Specifications
300°C (decomposition) | |
White | |
99.8-100.3% | |
MFCD00003534 | |
Soluble in water,sulfuric acid,nitric acid and acetic acid. | |
[Na+].[O-][I](=O)(=O)=O | |
213.89 | |
CHEBI:75226 | |
ACS | |
3.86 g/cm3 at 20°C |
7790-28-5 | |
Powder | |
INaO4 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
sodium;periodate | |
23667635 | |
213.89 | |
Odorless | |
Sodium metaperiodate, For analysis ACS |
Safety and Handling
P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
H271-H314-H335-H372
missing translation for 'dotInformation' : DOT Class: 5.1; Packaging Group: I
missing translation for 'einecsNumber' : 232-197-6
missing translation for 'rtecsNumber' : SD4550000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only