missing translation for 'onlineSavingsMsg'
Learn More

Sodium metasilicate nonahydrate, 44-47.5% total solids, Thermo Scientific Chemicals

£19.70 - £411.05

Chemical Identifiers

CAS 13517-24-3
Molecular Formula Na2SiO3·9H2O
Molecular Weight (g/mol) 284.19
MDL Number MFCD00149175
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
Synonym sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10607391
View Documents Promotion Details
Thermo Scientific Acros
419590250
25 g Plastic bottle
£19.70
25g
Please to purchase this item. Need a web account? Register with us today!
10045740
View Documents
Thermo Scientific Acros
419595000
500 g Plastic bottle
£126.63
500g
Please to purchase this item. Need a web account? Register with us today!
10388623
View Documents
Thermo Scientific Acros
419590025
2.5 kg Plastic bottle
£411.05
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13517-24-3
284.19
PHIQPXBZDGYJOG-UHFFFAOYSA-N
61639
O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
Na2SiO3·9H2O
MFCD00149175
sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
disodium;dioxido(oxo)silane;nonahydrate

Specifications

72.0°C
White
Na2SiO3·9H2O
MFCD00149175
sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
PHIQPXBZDGYJOG-UHFFFAOYSA-N
disodium;dioxido(oxo)silane;nonahydrate
61639
44 to 47.5% total solids
Sodium metasilicate nonahydrate, 44 - 47.5% total solids
13517-24-3
Granules or Granular Powder
Na2SiO3·9H2O
15, 8804
Solubility in water: soluble
O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
284.19
284.19
2.61
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

Product Identifier
  • Sodium metasilicate nonahydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only