missing translation for 'onlineSavingsMsg'
Learn More

Sodium metasilicate pentahydrate, tech.

Product Code. 11397697
Click to view available options
Quantity:
1 kg
5 x 1 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11397697

Brand: Thermo Scientific Alfa Aesar 014103.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10213-79-3 | H10Na2O8Si | 212.137 g/mol

Sodium metasilicate pentahydrate is used as a starting material in the preparation of silicas and zeolites. It finds application in detergents and cleaners, pulp and paper production. It is also used in soil stabilization, sealing, adhesives and binders. Further, it is used in titanium dioxide production, paints for masonry and glass, spray coatings for tunnel construction. In addition to this, it is used in fruit and vegetable washes and sanitizers for food-contact surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10213-79-3
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.137
MDL Number MFCD00149176
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate pentahydrate, silicic acid, disodium salt, pentahydrate, disodium oxosilanediolate pentahydrate, silicic acid h2sio3 , disodium salt, pentahydrate, 2na.so3i.5h2o, sodium meta-silicate pentahydrate, sodium silicate solution water glass, disodium dioxido oxo silane pentahydrate, sodium metasilicate pentahydrate, tech., disodium oxosilanebis olate pentahydrate
PubChem CID 61495
IUPAC Name disodium;dioxido(oxo)silane;pentahydrate
SMILES O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Melting Point 72°C
Color White
Physical Form Granules
Quantity 1 kg
UN Number UN3253
Merck Index 14,8642
Solubility Information Soluble in cold water.
Formula Weight 212.14 (122.06 Anhydrous)
Grade Technical
Odor Odorless
Density 1.75 g/mL
Chemical Name or Material Sodium metasilicate pentahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium metasilicate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium metasilicatepentahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.