missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium methanesulfonate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.
Solubility
Soluble in water, alcohol, ether, hexane, benzene, methylcyclopentane, toluene, o-chlorotoluene and ethyl disulfide.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.
Solubility
Soluble in water, alcohol, ether, hexane, benzene, methylcyclopentane, toluene, o-chlorotoluene and ethyl disulfide.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Odor | Odorless |
Linear Formula | CH3SO3Na |
Quantity | 5 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water,alcohol,ether,hexane,benzene,methylcyclopentane,toluene,o-chlorotoluene and ethyl disulfide. |
Formula Weight | 118.09 |
Percent Purity | 98% |
Chemical Name or Material | Sodium methanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'