missing translation for 'onlineSavingsMsg'
Learn More
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals Product Code.: 10347680

Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals

Product Code. 10347680
1 L
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
25 L
This item is not returnable. View return policy

Product Code. 10347680

Brand: Thermo Scientific Acros 168600010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4, 67-56-1
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755Show MoreShow Less
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Melting Point 1.0°C to 5.0°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 93.0°C
Flash Point 33°C
Packaging Glass Bottle
Linear Formula CH3ONa
Refractive Index 1.3701
Quantity 1 L
Beilstein 01,28
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Merck Index 15,8775
Specific Gravity 0.97
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol
Viscosity 68 mPa.s (20°C)
Formula Weight 54.02
Grade Pure
Physical Form Liquid
Chemical Name or Material Sodium methoxide, 30 wt% Solution in Methanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467897 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number168600025,168600100,168600010,168600250,168600251
Lot NumberA0458271 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number168600025,168600100,168600010,168600250,168600251
Lot NumberA0458272 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number168600025,168600100,168600010,168600250,168600251
Lot NumberA0465329 Certificate TypeCertificate of Analysis Date09/08/2024 Catalog Number168600025,168600100,168600010,168600250,168600251
Lot NumberA0462364 Certificate TypeCertificate of Analysis Date01/05/2024 Catalog Number168600025,168600100,168600010,168600250,168600251
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Methyl alcohol, Sodium methoxide
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.