missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, ACS Reagent, Honeywell Fluka™

Product Code. 15613890 Shop All Honeywell Fluka Products
2.5 kg
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
6 x 500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10102-40-6
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.93
MDL Number MFCD00149170
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdateShow MoreShow Less
PubChem CID 16211258
ChEBI CHEBI:75213
SMILES [Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Quantity 2.5 kg
Linear Formula Na2MoO4 · 2H2O
UN Number NONH for all modes of transport
IUPAC Name disodium;dioxido(dioxo)molybdenum;dihydrate
Formula Weight 241.95g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Sodium Molybdate Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Molybdate Dihydrate, ACS Reagent, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.