missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Molybdate Dihydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
Description
Sodium Molybdate Dihydrate, Extra Pure, SLR
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
pH | 9.0 to 10 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Formula Weight | 241.95g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'