missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Molybdate Dihydrate, puriss., Honeywell Fluka™
Puriss., meets analytical specification of Ph Eur, 99-100.5% (calc. to the dried substance)
£324.70 - £2187.06
Chemical Identifiers
CAS | 10102-40-6 |
---|---|
Molecular Formula | MoNa2O4 |
Molecular Weight (g/mol) | 205.93 |
MDL Number | MFCD00149170 |
InChI Key | TVXXNOYZHKPKGW-UHFFFAOYSA-N |
Synonym | sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate |
PubChem CID | 16211258 |
ChEBI | CHEBI:75213 |
SMILES | [Na+].[Na+].[O-][Mo]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15641290
|
Honeywell-Fluka
13315-1KG |
1 kg |
£324.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15651290
|
Honeywell-Fluka
13315-2.5KG |
2.5 kg |
£603.88
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15661290
|
Honeywell-Fluka
13315-6X2.5KG |
6 x 2.5 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10102-40-6 | |
205.93 | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
16211258 | |
[Na+].[Na+].[O-][Mo]([O-])(=O)=O |
MoNa2O4 | |
MFCD00149170 | |
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
CHEBI:75213 |
Specifications
10102-40-6 | |
Na2MoO4 · 2H2O | |
NONH for all modes of transport | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
disodium;dioxido(dioxo)molybdenum;dihydrate | |
16211258 | |
241.95g/mol | |
puriss. |
MoNa2O4 | |
MFCD00149170 | |
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
[Na+].[Na+].[O-][Mo]([O-])(=O)=O | |
205.93 | |
CHEBI:75213 | |
99 to 100.5% (calc. to the dried substance) | |
Sodium Molybdate Dihydrate, Meets analytical specification of PhEur |
Safety and Handling
missing translation for 'einecsNumber' : 231-551-7