missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium molybdate dihydrate, Puriss. p.a., ≥99.0% (T), Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Specifications
Melting Point | 100°C (decomposition)(literature) |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Quantity | 100 g |
Linear Formula | Na2MoO4 · 2H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 241.95 |
Percent Purity | >99% |
Grade | puriss. p.a. |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'