missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdate(VI) dihydrate, 99+%, Thermo Scientific Chemicals

£25.33 - £589.94

Chemical Identifiers

CAS 10102-40-6
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.93
MDL Number MFCD00149170
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium;dioxido(dioxo)molybdenum;dihydrate
SMILES [Na+].[Na+].[O-][Mo]([O-])(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10164830
View Documents Promotion Details
Thermo Scientific Acros
206370050
5 g Glass Bottle
£25.33
5g
Please to purchase this item. Need a web account? Register with us today!
10503381
View Documents Promotion Details
Thermo Scientific Acros
206371000
100 g Plastic bottle
£48.74
100g
Please to purchase this item. Need a web account? Register with us today!
10513381
View Documents Promotion Details
Thermo Scientific Acros
206375000
500 g Plastic bottle
£146.73
500g
Please to purchase this item. Need a web account? Register with us today!
10043113
View Documents Promotion Details
Thermo Scientific Acros
206370025
2.5 kg Plastic bottle
£589.94
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

10102-40-6
205.93
TVXXNOYZHKPKGW-UHFFFAOYSA-N
16211258
disodium;dioxido(dioxo)molybdenum;dihydrate
MoNa2O4
MFCD00149170
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
CHEBI:75213
[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

10102-40-6
Crystalline Powder
MoNa2O4
MFCD00149170
15, 8776
Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C)
[Na+].[Na+].[O-][Mo]([O-])(=O)=O
205.93
CHEBI:75213
≥99%
3.2800g/mL
White
99% min
Na2MoO4·2H2O
15,294
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
TVXXNOYZHKPKGW-UHFFFAOYSA-N
disodium;dioxido(dioxo)molybdenum;dihydrate
16211258
241.95
3.28
Sodium molybdate(VI) dihydrate, 99+%
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : QA5085000

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only