missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdenum oxide dihydrate, ACS, 99.5-103.0%, Thermo Scientific Chemicals

Product Code. 11306628
Click to view available options
Quantity:
100 g
500 g

It is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.93
MDL Number MFCD00149170
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium;dioxido(dioxo)molybdenum;dihydrate
SMILES [Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 100°C-2H2O
Color Green
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5-103.0%
Solubility Information Soluble in water.
Formula Weight 241.95 (205.92 Anhydrous)
Grade ACS
Odor Odorless
Appearance White powder/flake/crystalline/beads
Density 2.71
Chemical Name or Material Sodium molybdenum oxide dihydrate, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'