Learn More
Sodium molybdenum oxide dihydrate, ACS, 99.5-103.0%, Thermo Scientific Chemicals
£40.10 - £135.00
Chemical Identifiers
CAS | 10102-40-6 |
---|---|
Molecular Formula | MoNa2O4 |
Molecular Weight (g/mol) | 205.93 |
MDL Number | MFCD00149170 |
InChI Key | TVXXNOYZHKPKGW-UHFFFAOYSA-N |
Synonym | sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate |
PubChem CID | 16211258 |
ChEBI | CHEBI:75213 |
IUPAC Name | disodium;dioxido(dioxo)molybdenum;dihydrate |
SMILES | [Na+].[Na+].[O-][Mo]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306628
|
Thermo Scientific Alfa Aesar
012214.22 |
100 g |
£40.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396618
|
Thermo Scientific Alfa Aesar
012214.36 |
500 g |
£135.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10102-40-6 | |
205.93 | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
16211258 | |
disodium;dioxido(dioxo)molybdenum;dihydrate |
MoNa2O4 | |
MFCD00149170 | |
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
CHEBI:75213 | |
[Na+].[Na+].[O-][Mo]([O-])(=O)=O |
Specifications
100°C-2H2O | |
Green | |
99.5-103.0% | |
MFCD00149170 | |
Soluble in water. | |
[Na+].[Na+].[O-][Mo]([O-])(=O)=O | |
205.93 | |
CHEBI:75213 | |
ACS | |
White powder/flake/crystalline/beads | |
Sodium molybdenum oxide dihydrate, For analysis ACS |
10102-40-6 | |
Powder | |
MoNa2O4 | |
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
disodium;dioxido(dioxo)molybdenum;dihydrate | |
16211258 | |
241.95 (205.92 Anhydrous) | |
Odorless | |
2.71 |
Safety and Handling
missing translation for 'einecsNumber' : 231-551-7
missing translation for 'rtecsNumber' : QA5085000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only