missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-dodecyl sulfate (SDS), 20% aq. soln., Thermo Scientific Chemicals

£53.37 - £84.32

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465639
View Documents
Thermo Scientific Alfa Aesar
J63394.AK
250 mL
£53.37
250mL
Please to purchase this item. Need a web account? Register with us today!
15475639
View Documents
Thermo Scientific Alfa Aesar
J63394.AP
500 mL
£84.32
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.

Solubility
Miscible with ethanol.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
CHEBI:8984
C12H25NaO4S
MFCD00036175
3423265
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Colorless
Liquid
C12H25NaO4S
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium;dodecyl sulfate
3423265
288.38
151-21-3
20% aq. soln.
CH3(CH2)11OSO3Na
Miscible with ethanol.
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
Sodium n-dodecyl sulfate (SDS)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362

H315-H318

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Sodium n-dodecyl sulfate (SDS), 20% aq. soln.

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Sodium lauryl sulfate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only