missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
3 kg
Description
Sodium Nitrate, Extra Pure, SLR
Specifications
Specifications
Boiling Point | 380°C |
Melting Point | 306°C |
Color | White |
pH | 5.5 to 8 |
Physical Form | Solid Crystalline |
Quantity | 3 kg |
IUPAC Name | sodium nitrate |
Formula Weight | 84.99g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'