Learn More
Sodium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 308°C |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.999% (metals basis) |
Solubility Information | Freely soluble in water,ammonia and hydrazine; soluble in alcohol; slightly soluble in pyridine. |
Formula Weight | 84.99 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White crystals |
Density | 2.26 g/cm3 at 20°C |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X22K036 | Certificate of Analysis | 05/11/2024 | 010873.06,010873.14,010873.22 |
X02K019 | Certificate of Analysis | 09/10/2024 | 010873.06,010873.14,010873.22 |
X02J019 | Certificate of Analysis | 11/10/2023 | 010873.06,010873.14,010873.22 |
Y30I042 | Certificate of Analysis | 05/01/2023 | 010858.06,010858.14,010873.06,010858.22 |
X31I034 | Certificate of Analysis | 08/11/2022 | 010873.06,010926.30,010873.14,010873.22 |
Safety and Handling


- Sodium nitrate
- Warning
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- MIXTURE LIST-Contains : Sodium nitrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.