missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 g |
Linear Formula | NaNO3 |
UN Number | 1498 |
Formula Weight | 84.99g/mol |
Percent Purity | ≥99.5% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Sodium Nitrate, Reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'