missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite 0.5 M (0.5 N) Analytical Volumetric Solution (AVL), Reagecon
Fully factorized, high purity, stable product, developed and tested for titrations.
Brand: Reagecon NANO055
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
Specifications
Accuracy ± 0.1% | |
HDPE Container | |
Titration |
0.5 M (0.5 N) | |
5 L | |
Liquid |