missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite 0.5 M (0.5 N) Analytical Volumetric Solution (AVL), Reagecon

Fully factorized, high purity, stable product, developed and tested for titrations.

Brand:  Reagecon NANO051

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11572992

  • £46.35 / 1 L
Estimated Shipment: 26-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified to a specification of ± 0.1%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
Specifications

Specifications

Accuracy ± 0.1%
HDPE Bottle GL45
Titration
0.5 M (0.5 N)
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers