Learn More
Sodium Nitrite, +97%, For analysis, Thermo Scientific Chemicals
£231.98
Chemical Identifiers
CAS | 7632-00-0 |
---|---|
Molecular Formula | NNaO2 |
Molecular Weight (g/mol) | 69.00 |
MDL Number | MFCD00011118 |
InChI Key | LPXPTNMVRIOKMN-UHFFFAOYSA-M |
Synonym | sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts |
PubChem CID | 23668193 |
ChEBI | CHEBI:78870 |
IUPAC Name | sodium;nitrite |
SMILES | [Na+].[O-]N=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11974261
|
Thermo Scientific Acros
424350051 |
5 kg | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7632-00-0 | |
69.00 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
23668193 | |
sodium;nitrite |
NNaO2 | |
MFCD00011118 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
CHEBI:78870 | |
[Na+].[O-]N=O |
Specifications
7632-00-0 | |
Crystalline Powder and/or Crystals | |
NNaO2 | |
MFCD00011118 | |
15, 8779 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
sodium;nitrite | |
23668193 | |
69 | |
ACS Reagent | |
>320°C |
Yellow | |
97+% | |
NaNO2 | |
01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
[Na+].[O-]N=O | |
69.00 | |
CHEBI:78870 | |
≥97.0% | |
Glass bottle | |
Sodium nitrite |
Safety and Handling
GHS P Statement
Toxic if swallowed.
Very toxic to aquatic life.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 231-555-9
- Sodium nitrite
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 3
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only