missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Specifications
Specifications
Boiling Point | 320°C |
Melting Point | 271°C |
Color | Yellow |
pH | 8 to 9 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 69g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium nitrite
- Danger
- Oxidising solid Category 3
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'