missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitroprusside Dihydrate, puriss. p.a., ≥99%, ACS reagent, Honeywell Fluka™
6.22 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Use promo code "21640" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Quantity | 25 g |
Linear Formula | Na2[Fe(CN)5NO] · 2H2O |
UN Number | UN3288 |
Formula Weight | 297.95g/mol |
Percent Purity | ≥99% |
Chemical Name or Material | Sodium nitroprusside dihydrate |
Sicurezza e movimentazione
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
Identificativo prodotto
- Sodium nitroprusside dihydrate
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'