missing translation for 'onlineSavingsMsg'
Learn More

Sodium orthovanadate, 99%, Thermo Scientific Chemicals

£86.73 - £273.36

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.91
MDL Number MFCD00003511
InChI Key IHIXIJGXTJIKRB-UHFFFAOYSA-N
Synonym sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
PubChem CID 61671
ChEBI CHEBI:35607
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10114740
View Documents Promotion Details
Thermo Scientific Acros
205330500
50 g Plastic bottle
£86.73
50g
Please to purchase this item. Need a web account? Register with us today!
10388480
View Documents
Thermo Scientific Acros
205332500
250 g Plastic bottle
£273.36
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13721-39-6
183.91
IHIXIJGXTJIKRB-UHFFFAOYSA-N
61671
trisodium;trioxido(oxo)vanadium
Na3O4V
MFCD00003511
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
CHEBI:35607
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

850.0°C to 866.0°C
White
98.5% min. (as Na3VO4)
Na3VO4
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
IHIXIJGXTJIKRB-UHFFFAOYSA-N
trisodium;trioxido(oxo)vanadium
61671
183.91
Sodium orthovanadate, 99%
13721-39-6
Powder
Na3O4V
MFCD00003511
Solubility in water: soluble. Other solubilities: insoluble in ethanol
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]
183.91
CHEBI:35607
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 237-287-9

missing translation for 'rtecsNumber' : YW1120000

missing translation for 'tsca' : TSCA

Product Identifier
  • Sodium orthovanadate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only