missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium oxalate, ACS reagent, Thermo Scientific Chemicals
£33.00 - £111.00
Chemical Identifiers
CAS | 62-76-0 |
---|---|
Molecular Formula | C2Na2O4 |
Molecular Weight (g/mol) | 134.00 |
MDL Number | MFCD00012465 |
InChI Key | ZNCPFRVNHGOPAG-UHFFFAOYSA-L |
Synonym | sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt |
PubChem CID | 6125 |
IUPAC Name | disodium;oxalate |
SMILES | [Na+].[Na+].[O-]C(=O)C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12367183
|
Thermo Scientific Acros
446961000 |
100 g | Plastic bottle |
£33.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12377183
|
Thermo Scientific Acros
446965000 |
500 g | Plastic bottle |
£111.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62-76-0 | |
134.00 | |
ZNCPFRVNHGOPAG-UHFFFAOYSA-L | |
6125 | |
[Na+].[Na+].[O-]C(=O)C([O-])=O |
C2Na2O4 | |
MFCD00012465 | |
sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt | |
disodium;oxalate |
Specifications
250.0°C to 270.0°C | |
C2Na2O4 | |
MFCD00012465 | |
sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt | |
ZNCPFRVNHGOPAG-UHFFFAOYSA-L | |
disodium;oxalate | |
6125 | |
ACS Reagent |
62-76-0 | |
NaO2CCO2Na | |
15,8781 | |
Solubility in water: 37g/L (20°C). Other solubilities: 6.3 g/100mL water (100°C), insoluble in ethanol and ether | |
[Na+].[Na+].[O-]C(=O)C([O-])=O | |
134.00 | |
134 | |
Sodium oxalate |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-550-3
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium oxalate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only