Learn More
Sodium (para)periodate, 98%, Thermo Scientific Chemicals
£56.90 - £246.00
Chemical Identifiers
CAS | 13940-38-0 |
---|---|
Molecular Formula | H2INa3O6 |
Molecular Weight (g/mol) | 293.88 |
MDL Number | MFCD00036146 |
InChI Key | DSYMXPNUZLZKOA-UHFFFAOYSA-K |
Synonym | sodium paraperiodate, sodium para periodate, sodium triparaperiodate, sodium hydrogen periodate, 3na.h2io6, periodic acid trisodium salt, trisodium dihydrogen orthoperiodate, sodium para periodate, commercial, trisodium dihydroxydioxidoiodosylolate, trisodium dihydroxy-trioxido-oxo-$l^ 7-iodane |
PubChem CID | 9796123 |
IUPAC Name | trisodium;dihydroxy-trioxido-oxo-$l^{7}-iodane |
SMILES | [Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10677400
|
Thermo Scientific Acros
432851000 |
100 g | Glass bottle |
£56.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10422180
|
Thermo Scientific Acros
432855000 |
500 g | Glass bottle |
£246.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13940-38-0 | |
293.88 | |
DSYMXPNUZLZKOA-UHFFFAOYSA-K | |
9796123 | |
[Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O |
H2INa3O6 | |
MFCD00036146 | |
sodium paraperiodate, sodium para periodate, sodium triparaperiodate, sodium hydrogen periodate, 3na.h2io6, periodic acid trisodium salt, trisodium dihydrogen orthoperiodate, sodium para periodate, commercial, trisodium dihydroxydioxidoiodosylolate, trisodium dihydroxy-trioxido-oxo-$l^ 7-iodane | |
trisodium;dihydroxy-trioxido-oxo-$l^{7}-iodane |
Specifications
>300.0°C | |
White | |
97.5% min. (Iodometry) | |
Na3H2IO6 | |
sodium paraperiodate, sodium para periodate, sodium triparaperiodate, sodium hydrogen periodate, 3na.h2io6, periodic acid trisodium salt, trisodium dihydrogen orthoperiodate, sodium para periodate, commercial, trisodium dihydroxydioxidoiodosylolate, trisodium dihydroxy-trioxido-oxo-$l^ 7-iodane | |
DSYMXPNUZLZKOA-UHFFFAOYSA-K | |
trisodium;dihydroxy-trioxido-oxo-$l^{7}-iodane | |
9796123 | |
98% |
13940-38-0 | |
Powder | |
H2INa3O6 | |
MFCD00036146 | |
Solubility in water: slightly soluble | |
[Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O | |
293.88 | |
293.89 | |
Sodium (para)periodate |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the risk of explosion.
IF ON SKIN: Wash with plenty of soap an
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 237-720-1
- Sodium (para)periodate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only