missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate monohydrate, 95%, Thermo Scientific Chemicals

£23.62 - £307.53

Chemical Identifiers

CAS 10332-33-9
Molecular Formula BNaO3·H2O
Molecular Weight (g/mol) 99.81
MDL Number MFCD00149230
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10678624
View Documents
Thermo Scientific Acros
386410250
25 g Plastic bottle
£23.62
25g
Please to purchase this item. Need a web account? Register with us today!
10678814
View Documents
Thermo Scientific Acros
386412500
250 g Plastic bottle
£108.54
250g
Please to purchase this item. Need a web account? Register with us today!
10337273
View Documents
Thermo Scientific Acros
386410010
1 kg Plastic bottle
£307.53
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

10332-33-9
BNaO3·H2O
99.81
MFCD00149230

Specifications

10332-33-9
Crystalline Powder
BNaO3·H2O
Solubility in water: 15g/L (20°C)
99.81
Sodium perborate monohydrate
White
94% min. (active oxygen) (Iodometry)
MFCD00149230
99.81
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Obtain special instructions before use.
Avoid

GHS Signal Word: Danger

Product Identifier
  • Sodium perborate monohydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only