missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate tetrahydrate, 97%, pure, Thermo Scientific Chemicals

Product Code. 10613373
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10486-00-7
Molecular Formula BNaO3·4H2O
Molecular Weight (g/mol) 153.86

Specifications

Melting Point 60.0°C
Color White
pH 10.1 to 10.6 (1% soln.)
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 97%
Linear Formula NaBO3·4H2O
Fieser 01,1102; 02,387; 04,59; 14,290; 16,310
Merck Index 14,8652
Solubility Information Solubility in water: 21.5g/L (18°C)
Formula Weight 153.86
Percent Purity 97%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Sodium perborate tetrahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perborate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium perborate tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'