missing translation for 'onlineSavingsMsg'
Learn More

Sodium percarbonate, 13-14% active oxygen, Thermo Scientific Chemicals

Product Code. 10522252
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15630-89-4
Molecular Formula 1·5 H2O2
Molecular Weight (g/mol) 157.01
MDL Number MFCD00043204
Synonym Sodium carbonate hydrogen peroxide

Specifications

Color White
Physical Form Granular Powder
Quantity 2.5 kg
Linear Formula 1·5 H2O2
Solubility Information Solubility in water: 150g/L.
Formula Weight 157.01
Percent Purity ≥13% (available oxygen)
Packaging Plastic bottle
Chemical Name or Material Sodium percarbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.