Learn More
Sodium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals
£23.70 - £289.44
Chemical Identifiers
CAS | 7601-89-0 |
---|---|
InChI Key | BAZAXWOYCMUHIX-UHFFFAOYSA-M |
Synonym | sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian |
PubChem CID | 522606 |
IUPAC Name | sodium;perchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10032570
|
Thermo Scientific Acros
197120050 |
5 g | Glass Bottle | N/A | |||||
10752241
|
Thermo Scientific Acros
197122500 |
250 g | Plastic bottle |
£104.52
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10012560
|
Thermo Scientific Acros
197120010 |
1 kg | Plastic bottle |
£289.44
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7601-89-0 | |
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian | |
sodium;perchlorate |
BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
522606 | |
[O-]Cl(=O)(=O)=O.[Na+] |
Specifications
7601-89-0 | |
6.0 to 8.0 (5% aq. soln.) | |
99+% | |
NaClO4 | |
BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
sodium;perchlorate | |
522606 | |
≥99.0% (on dry substance) | |
2.0% max. (130°C, 2 hrs) |
White | |
Crystalline Powder and/or Crystals | |
ClNaO4 | |
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian | |
[O-]Cl(=O)(=O)=O.[Na+] | |
122.436 | |
122.44 | |
Analytical | |
Sodium perchlorate |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Sodium perchlorate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.