missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate Monohydrate, 98%, ACS Reagent, Honeywell Fluka™

Catalog Number 15693190
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7791-07-3
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7791-07-3

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 469°C
Quantity 100 g
Molecular Formula ClH2NaO5
Linear Formula NaClO4 · H2O
MDL Number MFCD00149164
UN Number UN1502
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]
IUPAC Name sodium;perchlorate;hydrate
Molecular Weight (g/mol) 140.45
PubChem CID 516899
Formula Weight 140.46g/mol
Percent Purity 98%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.02g/cm3
Chemical Name or Material Sodium Perchlorate Monohydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'